Product Details

SNP ID
rs77105320
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:231248084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTACAGTTGACCTTTGAACAAC[A/G]TGGGAGTTGGGGGCACCAACCCTCT
Phenotype
MIM: 602744
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf131 PubMed Links

Gene Details

Gene
C1orf131
Gene Name
chromosome 1 open reading frame 131
There are no transcripts associated with this gene.

Gene
GNPAT
Gene Name
glyceronephosphate O-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316350.1 Intron NP_001303279.1
NM_014236.3 Intron NP_055051.1
XM_005273313.4 Intron XP_005273370.1
XM_011544303.2 Intron XP_011542605.1
XM_011544304.2 Intron XP_011542606.1

View Full Product Details