Product Details

SNP ID
rs79240902
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.1:235548052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTCTGGGATTCCATCGTGTGAA[C/T]ATACCCTAATGTGCTTATCCAGCCA
Phenotype
MIM: 604388
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GNG4 PubMed Links

Gene Details

Gene
GNG4
Gene Name
G protein subunit gamma 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098721.1 4314 UTR 3 NP_001092191.1
NM_001098722.1 4314 UTR 3 NP_001092192.1
NM_004485.3 4314 UTR 3 NP_004476.1
XM_006711761.2 4314 UTR 3 XP_006711824.1
XM_011544167.2 4314 UTR 3 XP_011542469.1

View Full Product Details