Product Details

SNP ID
rs74541106
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:236142784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTCGTCTACACCGTGTTCTACGC[G/T]CTGCTCTTCGTGTTCATCTACGTGC
Phenotype
MIM: 604658
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR137B PubMed Links

Gene Details

Gene
GPR137B
Gene Name
G protein-coupled receptor 137B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003272.3 253 Silent Mutation GCG,GCT A54A NP_003263.1
XM_017002209.1 253 Silent Mutation GCG,GCT A54A XP_016857698.1
XM_017002210.1 253 Silent Mutation GCG,GCT A54A XP_016857699.1

View Full Product Details