Product Details

SNP ID
rs79542980
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:236689813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGTGTTGGTGTCTAGAAGTAAA[C/T]ATCTCCTGGAACCAAGGAACACATA
Phenotype
MIM: 102573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTN2 PubMed Links

Gene Details

Gene
ACTN2
Gene Name
actinin alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103.3 Intron NP_001094.1
NM_001278343.1 Intron NP_001265272.1
NM_001278344.1 Intron NP_001265273.1

View Full Product Details