Product Details

SNP ID
rs75083787
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:206639405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTGCCAATTGGTTCACCTCTA[C/T]AGGAAACTATTTGCAATTGGGTTGA
Phenotype
MIM: 603497
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYRK3 PubMed Links

Gene Details

Gene
DYRK3
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004023.1 Intron NP_001004023.1
NM_003582.2 Intron NP_003573.2
XM_005273315.4 Intron XP_005273372.1
XM_011510061.2 Intron XP_011508363.1

View Full Product Details