Product Details

SNP ID
rs79217065
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:247915023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTATGCAGGGGTTATGCAGAGT[A/G]TAGGGTCTGCGTGTCTTTTTGCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2T8 PubMed Links

Gene Details

Gene
OR2T8
Gene Name
olfactory receptor family 2 subfamily T member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005522.1 886 Intron NP_001005522.1
XM_011544178.2 886 Intron XP_011542480.1
XM_017001172.1 886 UTR 5 XP_016856661.1

View Full Product Details