Product Details

SNP ID
rs79016895
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:212047144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATTTCACATTTACCATTTTCTT[C/T]GTTTAGAATGGATGGCTCACTGGAA
Phenotype
MIM: 610617
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DTL PubMed Links

Gene Details

Gene
DTL
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286229.1 Intron NP_001273158.1
NM_001286230.1 Intron NP_001273159.1
NM_016448.3 Intron NP_057532.3
XM_011509614.1 Intron XP_011507916.1
Gene
RPL21P28
Gene Name
ribosomal protein L21 pseudogene 28
There are no transcripts associated with this gene.

View Full Product Details