Product Details

SNP ID
rs76240098
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:212688126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAACACTCTCACTTTCAAATGTA[A/C]CTGTTGGGTTTGGTTCCATTTAATA
Phenotype
MIM: 612470
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BATF3 PubMed Links

Gene Details

Gene
BATF3
Gene Name
basic leucine zipper ATF-like transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018664.2 Intron NP_061134.1
XM_017001683.1 Intron XP_016857172.1

View Full Product Details