Product Details

SNP ID
rs78889390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27871443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGAGCAGAGGCAGAGTAAAGGG[A/G]GGAAATTCATCACTTGACGTTGCCT
Phenotype
MIM: 611306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCARA5 PubMed Links

Gene Details

Gene
SCARA5
Gene Name
scavenger receptor class A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173833.5 2590 UTR 3 NP_776194.2

View Full Product Details