Product Details

SNP ID
rs77766600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37861371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCTGTTTTCTACTGACTTTTAA[C/T]TTTTATTAACTACATTAAGCCCTTC
Phenotype
MIM: 608737
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB11FIP1 PubMed Links

Gene Details

Gene
RAB11FIP1
Gene Name
RAB11 family interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002814.2 5432 UTR 3 NP_001002814.2
NM_025151.4 5432 UTR 3 NP_079427.4
XM_017013869.1 5432 Intron XP_016869358.1

View Full Product Details