Product Details

SNP ID
rs78137824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:17644116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGGGGATGAATGGCAAACCCAA[C/G]TTTGGCTAATGTCATTGAGAACAAC
Phenotype
MIM: 609589 MIM: 604584
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MTUS1 PubMed Links

Gene Details

Gene
MTUS1
Gene Name
microtubule associated tumor suppressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001924.2 3003 UTR 3 NP_001001924.1
NM_001001925.2 3003 UTR 3 NP_001001925.1
NM_001001931.2 3003 UTR 3 NP_001001931.1
NM_001166393.1 3003 UTR 3 NP_001159865.1
NM_020749.4 3003 UTR 3 NP_065800.1
XM_005273577.3 3003 UTR 3 XP_005273634.1
XM_005273578.3 3003 UTR 3 XP_005273635.1
XM_005273579.3 3003 UTR 3 XP_005273636.1
XM_005273580.3 3003 UTR 3 XP_005273637.1
XM_005273581.3 3003 UTR 3 XP_005273638.1
XM_005273582.3 3003 UTR 3 XP_005273639.1
XM_005273583.4 3003 Intron XP_005273640.1
XM_011544595.2 3003 UTR 3 XP_011542897.1
XM_017013698.1 3003 UTR 3 XP_016869187.1
XM_017013699.1 3003 UTR 3 XP_016869188.1
XM_017013700.1 3003 UTR 3 XP_016869189.1
XM_017013701.1 3003 UTR 3 XP_016869190.1
XM_017013702.1 3003 UTR 3 XP_016869191.1
XM_017013703.1 3003 Intron XP_016869192.1
XM_017013704.1 3003 UTR 3 XP_016869193.1
Gene
PDGFRL
Gene Name
platelet derived growth factor receptor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006207.2 3003 Intron NP_006198.1

View Full Product Details