Product Details

SNP ID
rs76893263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:18527741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAGACCACCACAAACGCCATC[A/G]TTTTCTAGAAGACGGCATTTCCAGA
Phenotype
MIM: 614440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSD3 PubMed Links

Gene Details

Gene
PSD3
Gene Name
pleckstrin and Sec7 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015310.3 12536 UTR 3 NP_056125.3
NM_206909.2 12536 UTR 3 NP_996792.1
XM_011544467.2 12536 Intron XP_011542769.1
XM_011544468.2 12536 Intron XP_011542770.1
XM_011544469.2 12536 Intron XP_011542771.1
XM_011544471.2 12536 Intron XP_011542773.1
XM_011544473.2 12536 Intron XP_011542775.1
XM_017013258.1 12536 UTR 3 XP_016868747.1
XM_017013259.1 12536 UTR 3 XP_016868748.1
XM_017013260.1 12536 UTR 3 XP_016868749.1
XM_017013261.1 12536 UTR 3 XP_016868750.1
XM_017013262.1 12536 Intron XP_016868751.1
XM_017013263.1 12536 Intron XP_016868752.1
XM_017013264.1 12536 Intron XP_016868753.1
XM_017013265.1 12536 UTR 3 XP_016868754.1

View Full Product Details