Product Details

SNP ID
rs79320058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65540381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTCGTCCAGGCACTCGTCCAC[A/G]TCTGCAGGGAGGAAAAGCGTGGGTA
Phenotype
MIM: 602090 MIM: 607982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LTBP3 PubMed Links

Gene Details

Gene
LTBP3
Gene Name
latent transforming growth factor beta binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130144.2 3559 Silent Mutation GAC,GAT D1036D NP_001123616.1
NM_001164266.1 3559 Silent Mutation GAC,GAT D919D NP_001157738.1
NM_021070.4 3559 Silent Mutation GAC,GAT D1036D NP_066548.2
XM_011545032.2 3559 Silent Mutation GAC,GAT D1045D XP_011543334.1
XM_011545033.2 3559 Silent Mutation GAC,GAT D1045D XP_011543335.1
XM_017017737.1 3559 Intron XP_016873226.1
Gene
SCYL1
Gene Name
SCY1 like pseudokinase 1
There are no transcripts associated with this gene.

View Full Product Details