Product Details
- SNP ID
-
rs78789510
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:66332520 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCTGGGTCTCAGGCCACTCCGCC[C/T]GGCGGTAGACGAGGTAGCCAGTGGT
- Phenotype
-
MIM: 606259
MIM: 605965
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BRMS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1784029] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BRMS1
- Gene Name
- breast cancer metastasis suppressor 1
There are no transcripts associated with this gene.
- Gene
- RIN1
- Gene Name
- Ras and Rab interactor 1
View Full Product Details