Product Details

SNP ID
rs75898802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66547132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCAAAACCGAGGCCTGAGAGGC[A/C]GGGCTGTTTGTCCTGGGCATCTCAG
Phenotype
MIM: 102574
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACTN3 PubMed Links

Gene Details

Gene
ACTN3
Gene Name
actinin alpha 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104.3 Intron NP_001095.2
NM_001258371.2 Intron NP_001245300.2
Gene
ZDHHC24
Gene Name
zinc finger DHHC-type containing 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207340.1 Intron NP_997223.1
XM_005273874.3 Intron XP_005273931.1
XM_011544893.2 Intron XP_011543195.1
XM_011544894.1 Intron XP_011543196.1

View Full Product Details