Product Details

SNP ID
rs80257081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68066452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTAATTGTAACTGCTGTATTC[A/G]AAATCAATGAGCATCAGTTTCTGTT
Phenotype
MIM: 118491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHKA PubMed Links

Gene Details

Gene
CHKA
Gene Name
choline kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277.2 1091 Silent Mutation TTC,TTT F331F NP_001268.2
NM_212469.1 1091 Silent Mutation TTC,TTT F313F NP_997634.1
XM_017017147.1 1091 Silent Mutation TTC,TTT F293F XP_016872636.1
XM_017017148.1 1091 Silent Mutation TTC,TTT F155F XP_016872637.1

View Full Product Details