Product Details

SNP ID
rs76390346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56375774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGTTTGATCCTACTCATTAGA[A/G]CGGATACAAGTCTCAACACACCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8U1 PubMed Links

Gene Details

Gene
OR8U1
Gene Name
olfactory receptor family 8 subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005204.1 151 Missense Mutation ACG,GCG T51A NP_001005204.1

View Full Product Details