Product Details
- SNP ID
-
rs74603381
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:56375895 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAATTTCTTGTACAAACAAAATG[G/T]TATATCCTTTGATGCATGTGCTACT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR8U1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs559359089] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR8U1
- Gene Name
- olfactory receptor family 8 subfamily U member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005204.1 |
272 |
Missense Mutation |
GGT,GTT |
G91V |
NP_001005204.1 |
View Full Product Details