Product Details

SNP ID
rs80239514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72192974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGACCTGAGGTCTCCCTATAAC[A/G]TTTATTTTGCTACCACCCCTTTAAT
Phenotype
MIM: 136430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLR1 PubMed Links

Gene Details

Gene
FOLR1
Gene Name
folate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000802.3 Intron NP_000793.1
NM_016724.2 Intron NP_057936.1
NM_016725.2 Intron NP_057937.1
NM_016729.2 Intron NP_057941.1

View Full Product Details