Product Details

SNP ID
rs76191655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72195394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAGATGGCACCTGCCTGCAAA[C/T]GGCATTTCATCCAGGACACCTGCCT
Phenotype
MIM: 136430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOLR1 PubMed Links

Gene Details

Gene
FOLR1
Gene Name
folate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000802.3 337 Silent Mutation CGG,TGG R98W NP_000793.1
NM_016724.2 337 Silent Mutation CGG,TGG R98W NP_057936.1
NM_016725.2 337 Silent Mutation CGG,TGG R98W NP_057937.1
NM_016729.2 337 Silent Mutation CGG,TGG R98W NP_057941.1

View Full Product Details