Product Details

SNP ID
rs78465712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73309588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGTGATGGGTTAAATCTAAGCA[C/G]CATGAACTCAGCAGGGGTTTCTGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF17 PubMed Links

Gene Details

Gene
ARHGEF17
Gene Name
Rho guanine nucleotide exchange factor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014786.3 971 Missense Mutation ACC,AGC T317S NP_055601.2
XM_017018623.1 971 Intron XP_016874112.1
XM_017018624.1 971 Intron XP_016874113.1

View Full Product Details