Product Details

SNP ID
rs77638973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73950984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAACTCCCAGAGTGCACCTGTGC[A/G]GTTGTCAGGAGCAACCAAGGAAGCC
Phenotype
MIM: 610263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153614.3 364 UTR 5 NP_705842.2
XM_005273984.3 364 UTR 5 XP_005274041.1
XM_011545004.2 364 UTR 5 XP_011543306.1
XM_011545005.2 364 UTR 5 XP_011543307.1
XM_011545007.2 364 Intron XP_011543309.1
XM_011545009.2 364 UTR 5 XP_011543311.1
XM_011545013.2 364 UTR 5 XP_011543315.1
XM_011545014.2 364 UTR 5 XP_011543316.1
XM_011545015.2 364 Intron XP_011543317.1
XM_017017675.1 364 Intron XP_016873164.1
XM_017017676.1 364 Intron XP_016873165.1
XM_017017677.1 364 Intron XP_016873166.1
XM_017017678.1 364 Intron XP_016873167.1
XM_017017679.1 364 UTR 5 XP_016873168.1

View Full Product Details