Product Details

SNP ID
rs76629964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74001419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCCTTCTTGTCTTGTTCAAAAC[A/G]GTGATTCCCGTAACATCTGGACTTT
Phenotype
MIM: 602044
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UCP3 PubMed Links

Gene Details

Gene
UCP3
Gene Name
uncoupling protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003356.3 1287 Missense Mutation CCG,CTG P311L NP_003347.1
NM_022803.2 1287 Intron NP_073714.1

View Full Product Details