Product Details

SNP ID
rs79530724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74003092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTCAAGGGGCTGTTGGGATAAC[A/G]TGTGTAAAGTGCTCAGCACGAGGCC
Phenotype
MIM: 615944 MIM: 602044
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD3 PubMed Links

Gene Details

Gene
C2CD3
Gene Name
C2 calcium dependent domain containing 3
There are no transcripts associated with this gene.

Gene
UCP3
Gene Name
uncoupling protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003356.3 1914 Intron NP_003347.1
NM_022803.2 1914 UTR 3 NP_073714.1

View Full Product Details