Product Details

SNP ID
rs79279804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85660085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAACTTTACATTAACTTTTATA[C/T]TCTACTCAGCAGGGTGTCTGTCTGT
Phenotype
MIM: 606444 MIM: 612988
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREBZF PubMed Links

Gene Details

Gene
CREBZF
Gene Name
CREB/ATF bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039618.2 2322 UTR 3 NP_001034707.1
XM_006718642.3 2322 UTR 3 XP_006718705.1
XM_011545195.2 2322 UTR 3 XP_011543497.1
XM_017018086.1 2322 UTR 3 XP_016873575.1
XM_017018087.1 2322 UTR 3 XP_016873576.1
XM_017018088.1 2322 UTR 3 XP_016873577.1
XM_017018089.1 2322 UTR 3 XP_016873578.1
XM_017018090.1 2322 UTR 3 XP_016873579.1
XM_017018091.1 2322 UTR 3 XP_016873580.1
XM_017018092.1 2322 UTR 3 XP_016873581.1
Gene
TMEM126A
Gene Name
transmembrane protein 126A
There are no transcripts associated with this gene.

View Full Product Details