Product Details

SNP ID
rs79941863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58028263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTACCCTCCACCGTGGTTCCTAG[G/T]TCAGAGCAGGTGCCCTCTGCATTTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6Q1 PubMed Links

Gene Details

Gene
OR6Q1
Gene Name
olfactory receptor family 6 subfamily Q member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005212.3 Intron NP_001005212.1

View Full Product Details