Product Details

SNP ID
rs78218902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58118518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGATCTACATGCTCAGGGACAC[C/G]TGGAGTCTCCTAGCGACCTTTCTGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR9I1 PubMed Links

Gene Details

Gene
OR9I1
Gene Name
olfactory receptor family 9 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005211.1 927 Missense Mutation CAC,CAG H309Q NP_001005211.1
Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005212.3 927 Intron NP_001005212.1

View Full Product Details