Product Details

SNP ID
rs77034589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:90132668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCACACATAAAAATAATTCTGA[A/G]TGCTCTTGGAAAATGTTTCTTAAGC
Phenotype
MIM: 611636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAALAD2 PubMed Links

Gene Details

Gene
NAALAD2
Gene Name
N-acetylated alpha-linked acidic dipeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300930.1 Intron NP_001287859.1
NM_005467.3 Intron NP_005458.1
XM_017017043.1 Intron XP_016872532.1
XM_017017044.1 Intron XP_016872533.1
XM_017017045.1 Intron XP_016872534.1
XM_017017046.1 Intron XP_016872535.1

View Full Product Details