Product Details

SNP ID
rs80329505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95092689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCAAGTCAGCCAGGATCCAGA[A/G]AGTATTGGGAGAGAGGGAGGGAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENDOD1 PubMed Links

Gene Details

Gene
ENDOD1
Gene Name
endonuclease domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015036.2 Intron NP_055851.1

View Full Product Details