Product Details

SNP ID
rs75457182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59364943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTGCTTCTTTATTCCAAATTG[C/G]TGCTTTGCTTCAACTCCACTTCTGT
Phenotype
MIM: 615702
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5AN1 PubMed Links

Gene Details

Gene
OR5AN1
Gene Name
olfactory receptor family 5 subfamily AN member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004729.1 485 Missense Mutation GCT,GGT A162G NP_001004729.1

View Full Product Details