Product Details

SNP ID
rs78757629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59515152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTTCCTCCATCACAGCTCCTAA[A/G]GTCCTGATAGATCTTCTATCAGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D9 PubMed Links

Gene Details

Gene
OR4D9
Gene Name
olfactory receptor family 4 subfamily D member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004711.1 240 Silent Mutation AAA,AAG K80K NP_001004711.1

View Full Product Details