Product Details

SNP ID
rs76232633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61806692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAAGTATTTGTGCTGGTGGTT[G/T]TACGGCATATATTTTTTCTTCTGTT
Phenotype
MIM: 606148 MIM: 606149 MIM: 600393
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013402.4 750 Nonsense Mutation TAA,TAC *316Y NP_037534.3
XM_011545022.2 750 Nonsense Mutation TAA,TAC *245Y XP_011543324.1
Gene
FADS2
Gene Name
fatty acid desaturase 2
There are no transcripts associated with this gene.

Gene
FEN1
Gene Name
flap structure-specific endonuclease 1
There are no transcripts associated with this gene.

Gene
MIR1908
Gene Name
microRNA 1908
There are no transcripts associated with this gene.

View Full Product Details