Product Details

SNP ID
rs79273752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63540120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCATTCCATAGGTCCTGGCAGAA[C/T]GGAAGAAGAGACCGTTGCCTCCCGC
Phenotype
MIM: 605092
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RARRES3 PubMed Links

Gene Details

Gene
RARRES3
Gene Name
retinoic acid receptor responder 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004585.4 Intron NP_004576.2

View Full Product Details