Product Details

SNP ID
rs76877914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63553049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCTGCTGCCACACCTACTGTC[C/G]TGACTGCACCAGTGACCTGCAGCAG
Phenotype
MIM: 613866 MIM: 605092
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HRASLS2 PubMed Links

Gene Details

Gene
HRASLS2
Gene Name
HRAS like suppressor 2
There are no transcripts associated with this gene.

Gene
RARRES3
Gene Name
retinoic acid receptor responder 3
There are no transcripts associated with this gene.

View Full Product Details