Product Details

SNP ID
rs77887807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63760678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCTAACCTGTGAGGCCATAACTT[C/T]CATCAAAGCAGGAATCAATGGAATC
Phenotype
MIM: 615699 MIM: 604249
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf95 PubMed Links

Gene Details

Gene
C11orf95
Gene Name
chromosome 11 open reading frame 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144936.1 4796 UTR 3 NP_001138408.1
Gene
RTN3
Gene Name
reticulon 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265589.1 4796 Intron NP_001252518.1
NM_001265590.1 4796 Intron NP_001252519.1
NM_001265591.1 4796 Intron NP_001252520.1
NM_006054.3 4796 Intron NP_006045.1
NM_201428.2 4796 Intron NP_958831.1
NM_201429.2 4796 Intron NP_958832.1
NM_201430.2 4796 Intron NP_958833.1
XM_011544730.2 4796 Intron XP_011543032.1
XM_011544731.2 4796 Intron XP_011543033.1
XM_017017091.1 4796 Intron XP_016872580.1
XM_017017092.1 4796 Intron XP_016872581.1

View Full Product Details