Product Details

SNP ID
rs75184779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63767461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCTGTGACCTTTGATAGGTCA[C/T]GTCCGCACTCTGGGCCTGAGCTTCT
Phenotype
MIM: 615699 MIM: 604249
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf95 PubMed Links

Gene Details

Gene
C11orf95
Gene Name
chromosome 11 open reading frame 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144936.1 Intron NP_001138408.1
Gene
RTN3
Gene Name
reticulon 3
There are no transcripts associated with this gene.

View Full Product Details