Product Details

SNP ID
rs78646579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15803730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTTGCTAAGAGAGCAGAAGGC[A/C]CAAGAAGAATTCAGAATAAAGAAGG
Phenotype
MIM: 300028
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZRSR2 PubMed Links

Gene Details

Gene
ZRSR2
Gene Name
zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005089.3 1008 Intron NP_005080.1
XM_005274597.2 1008 UTR 5 XP_005274654.2
XM_011545589.2 1008 Silent Mutation GCA,GCC A56A XP_011543891.2
XM_011545590.2 1008 UTR 5 XP_011543892.2
XM_017029881.1 1008 Intron XP_016885370.1
XM_017029882.1 1008 UTR 5 XP_016885371.1
XM_017029883.1 1008 UTR 5 XP_016885372.1

View Full Product Details