Product Details

SNP ID
rs115321601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10726528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGACCTTGGATTCCCTGGTTGG[C/T]CTCTCTGATGAGTTGGGGAAACTCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1C2 PubMed Links

Gene Details

Gene
ATP6V1C2
Gene Name
ATPase H+ transporting V1 subunit C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039362.1 265 Silent Mutation GGC,GGT G52G NP_001034451.1
NM_144583.3 265 Silent Mutation GGC,GGT G52G NP_653184.2
XM_011510339.2 265 Silent Mutation GGC,GGT G52G XP_011508641.1
XM_011510340.2 265 Silent Mutation GGC,GGT G52G XP_011508642.1
XM_011510341.2 265 Silent Mutation GGC,GGT G52G XP_011508643.1
XM_017003745.1 265 Silent Mutation GGC,GGT G52G XP_016859234.1

View Full Product Details