Product Details

SNP ID
rs115462598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165750544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATCCATTCTCACTTCCGTGTA[A/C]TCACTTCTGTTACTCAGACTCTCAG
Phenotype
MIM: 601756
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GALNT3 PubMed Links

Gene Details

Gene
GALNT3
Gene Name
polypeptide N-acetylgalactosaminyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004482.3 Intron NP_004473.2
XM_005246449.1 Intron XP_005246506.1
XM_011510929.1 Intron XP_011509231.1
XM_017003770.1 Intron XP_016859259.1

View Full Product Details