Product Details

SNP ID
rs114841233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176117597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAAGGTCGGTATGAGCAGAGTT[A/G]CCACCCCAGCGGGGCGCGCAGCCCG
Phenotype
MIM: 142984 MIM: 142986 MIM: 142982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXD10 PubMed Links

Gene Details

Gene
HOXD10
Gene Name
homeobox D10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002148.3 Intron NP_002139.2
Gene
HOXD11
Gene Name
homeobox D11
There are no transcripts associated with this gene.

Gene
HOXD9
Gene Name
homeobox D9
There are no transcripts associated with this gene.

Gene
LOC100129455
Gene Name
uncharacterized LOC100129455
There are no transcripts associated with this gene.

View Full Product Details