Product Details

SNP ID
rs121434410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178436264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGTAAGTTGATCTTTTCACCA[G/A]GAGAATTCCTCAAGGAATGCCAAGT
Phenotype
MIM: 603424
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LOC101927027 PubMed Links

Gene Details

Gene
LOC101927027
Gene Name
uncharacterized LOC101927027
There are no transcripts associated with this gene.

Gene
PRKRA
Gene Name
protein activator of interferon induced protein kinase EIF2AK2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139517.1 907 Missense Mutation CCT,CTT P211L NP_001132989.1
NM_001139518.1 907 Missense Mutation CCT,CTT P197L NP_001132990.1
NM_001316362.1 907 Missense Mutation CCT,CTT P109L NP_001303291.1
NM_003690.4 907 Missense Mutation CCT,CTT P222L NP_003681.1
XM_011512063.1 907 Missense Mutation CCT,CTT P137L XP_011510365.1
XM_011512066.1 907 Missense Mutation CCT,CTT P109L XP_011510368.1
XM_017005159.1 907 Missense Mutation CCT,CTT P109L XP_016860648.1

View Full Product Details