Product Details

SNP ID
rs115207271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:181678123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGCGCTGCGCTGTAGGCGTG[C/T]GGCGGAGGCTTAACGTGGAAGACAT
Phenotype
MIM: 601724
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEUROD1 PubMed Links

Gene Details

Gene
NEUROD1
Gene Name
neuronal differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002500.4 985 Silent Mutation CCA,CCG P246P NP_002491.2

View Full Product Details