Product Details

SNP ID
rs115497017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:188735518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCATAGATGTATCATATCACCA[C/T]CCTTAAGCTTTGCACCTGGGTTGGC
Phenotype
MIM: 606423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIRC1 PubMed Links

Gene Details

Gene
DIRC1
Gene Name
disrupted in renal carcinoma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052952.2 Intron NP_443184.1
XM_017003281.1 Intron XP_016858770.1

View Full Product Details