Product Details

SNP ID
rs111450443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196135254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAAACATAATCATTTCATTAT[C/T]CAATGACTTACTAAACTTTACTTGC
Phenotype
MIM: 604727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK17B PubMed Links

Gene Details

Gene
STK17B
Gene Name
serine/threonine kinase 17b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004226.3 3636 UTR 3 NP_004217.1
XM_011512168.2 3636 UTR 3 XP_011510470.1
XM_011512169.1 3636 UTR 3 XP_011510471.1
XM_011512170.1 3636 UTR 3 XP_011510472.1
XM_011512171.2 3636 UTR 3 XP_011510473.1

View Full Product Details