Product Details

SNP ID
rs116150295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203713548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATAGCATTAAACATAAGAGGGA[A/G]AGGAGTGAATAATGGCTTGGGATTT
Phenotype
MIM: 186760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD28 PubMed Links

Gene Details

Gene
CD28
Gene Name
CD28 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243077.1 Intron NP_001230006.1
NM_001243078.1 Intron NP_001230007.1
NM_006139.3 Intron NP_006130.1
XM_011512194.2 Intron XP_011510496.1
XM_011512195.2 Intron XP_011510497.1
XM_011512197.2 Intron XP_011510499.1

View Full Product Details