Product Details

SNP ID
rs113252472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207539855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAATGGACAAAAAAATGAAAA[C/T]ATGGACATATAATGAACTTGACATA
Phenotype
MIM: 123810
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREB1 PubMed Links

Gene Details

Gene
CREB1
Gene Name
cAMP responsive element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320793.1 Intron NP_001307722.1
NM_004379.4 Intron NP_004370.1
NM_134442.4 Intron NP_604391.1
XM_011510645.1 Intron XP_011508947.1
XM_011510646.2 Intron XP_011508948.1
XM_011510647.2 Intron XP_011508949.1
XM_011510648.2 Intron XP_011508950.1
XM_011510649.2 Intron XP_011508951.1
XM_011510650.2 Intron XP_011508952.1
XM_011510651.2 Intron XP_011508953.1
XM_017003399.1 Intron XP_016858888.1
XM_017003400.1 Intron XP_016858889.1
XM_017003401.1 Intron XP_016858890.1

View Full Product Details