Product Details

SNP ID
rs111309414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216499928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATACTTACTTGGTTCATAGTG[A/C]AAATTGGTTCTCTTTTATAGACCAA
Phenotype
MIM: 613314
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPL37A PubMed Links

Gene Details

Gene
RPL37A
Gene Name
ribosomal protein L37a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000998.4 Intron NP_000989.1

View Full Product Details