Product Details
- SNP ID
-
rs114581128
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:217801700 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATACTGCAGTGCGGAGTGTTGTGT[A/G]TGCGTAGGTGTGTGCATGGGTGCAC
- Phenotype
-
MIM: 600076
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TNS1
PubMed Links
Gene Details
- Gene
- TNS1
- Gene Name
- tensin 1
View Full Product Details