Product Details

SNP ID
rs114000641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219259705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCTTCCCAGGGTCTCACCTTTA[C/T]CTTGGAGCAGGGCAGCAAGGTACTC
Phenotype
MIM: 604719 MIM: 191110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK16 PubMed Links

Gene Details

Gene
STK16
Gene Name
serine/threonine kinase 16
There are no transcripts associated with this gene.

Gene
TUBA4A
Gene Name
tubulin alpha 4a
There are no transcripts associated with this gene.

Gene
TUBA4B
Gene Name
tubulin alpha 4b
There are no transcripts associated with this gene.

View Full Product Details