Product Details

SNP ID
rs114600258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240140390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCAGGATGAACCCAGGAAGGGC[A/G]AGACCACCCATCCGTCAGGGCCGGC
Phenotype
MIM: 607877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPS9 PubMed Links

Gene Details

Gene
COPS9
Gene Name
COP9 signalosome subunit 9
There are no transcripts associated with this gene.

Gene
OTOS
Gene Name
otospiralin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_148961.3 92 UTR 5 NP_683764.1
XM_017003409.1 92 Intron XP_016858898.1
XM_017003410.1 92 Intron XP_016858899.1

View Full Product Details